missing translation for 'onlineSavingsMsg'
Learn More

Niraparib, Mikromol™

Product Code. 30052191
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052191 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052191 Supplier Mikromol Supplier No. MM3944.000250
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Niraparib
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Niraparib
CAS 1038915-60-4
Analyte or Component Names Niraparib
Shipping Condition Ice pack (-20°C)
Molecular Formula C19 H20 N4 O
Synonym 2H-Indazole-7-carboxamide, 2-[4-(3S)-3-piperidinylphenyl]-, 2-[4-(3S)-3-Piperidinylphenyl]-2H-indazole-7-carboxamide, Niraparib, Zejula, MK-4827
InChI Formula InChI=1S/C19H20N4O/c20-19(24)17-5-1-3-15-12-23(22-18(15)17)16-8-6-13(7-9-16)14-4-2-10-21-11-14/h1,3,5-9,12,14,21H,2,4,10-11H2,(H2,20,24)/t14-/m1/s1
SMILES NC(=O)c1cccc2cn(nc12)c3ccc(cc3)[C@@H]4CCCNC4
IUPAC Name 2-[4-[(3S)-3-piperidyl]phenyl]indazole-7-carboxamide
Molecular Weight (g/mol) 320.39
Quantity 250 mg
Formula Weight 320.1637 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.