missing translation for 'onlineSavingsMsg'
Learn More

Niraparib, TRC

Product Code. 30137331
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137331 5 mg 5mg
17940555 2.5 mg 2.5mg
17984985 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137331 Supplier Toronto Research Chemicals Supplier No. TRCN4814005MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1038915-60-4
Molecular Formula C19 H20 N4 O
Molecular Weight (g/mol) 320.39
Synonym 2H-Indazole-7-carboxamide, 2-[4-(3S)-3-piperidinylphenyl]-, 2-[4-(3S)-3-Piperidinylphenyl]-2H-indazole-7-carboxamide, Niraparib, Zejula
IUPAC Name 2-[4-[(3S)-piperidin-3-yl]phenyl]indazole-7-carboxamide
SMILES NC(=O)c1cccc2cn(nc12)c3ccc(cc3)[C@@H]4CCCNC4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H20N4O/c20-19(24)17-5-1-3-15-12-23(22-18(15)17)16-8-6-13(7-9-16)14-4-2-10-21-11-14/h1,3,5-9,12,14,21H,2,4,10-11H2,(H2,20,24)/t14-/m1/s1
Formula Weight 320.1637
Chemical Name or Material Niraparib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.