missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitazoxanide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Nitazoxanide
Specifications
Specifications
| Chemical Name or Material | Nitazoxanide |
| CAS | 55981-09-4 |
| Analyte or Component Names | Nitazoxanide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H9 N3 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzamide, 2-(acetyloxy)-N-(5-nitro-2-thiazolyl)-, Alinia, BRN 1225475, Cryptaz, DRG 0242, Daxon, Dexidex, Kidonax, Mitafar, N-(5-Nitro-2-thiazolyl)salicylamide acetate (ester), NSC 697855, NTX, NTZ, Nitacure, Nitacure 500, Nitarid, Nitazode, Nitazox 500, Nitazoxanida, Nitazoxanide, Nixide, Nixoran, Nizonide, PH 5776, Pacovanton, ParaMix, Xerovirin-C |
| InChI Formula | InChI=1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17) |
| SMILES | CC(=O)Oc1ccccc1C(=O)Nc2ncc(s2)[N+](=O)[O-] |
| IUPAC Name | [2-[(5-nitro-1,3-thiazol-2-yl)carbamoyl]phenyl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction