Learn More
Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.27in), Alloy M, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Nitinol compression coil spring, Alloy M |
| Name Note | Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.) |
| Quantity | 100 Pc. |
| Odor | Odorless |
| MDL Number | MFCD02091734 |