Learn More
Nitrendipine, LoGiCal
Product Code. 30463375
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463375 Supplier LoGiCal Supplier No. LGCFOR0092.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nitrendipine
CAS 39562-70-4
Analyte or Component Names Nitrendipine
Shipping Condition Room Temperature
Molecular Formula C18 H20 N2 O6
Certifications/Compliance ISO 9001
Synonym Nitrendipine, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid ethyl methyl ester, BAY-e 5009, Bayotensin, Baypress, Bylotensin, Deiten, Nidrel, Nitrendipin, Nitrepin, (+/-)-BAY E 5009, (+/-)-Nitrendipine
InChI Formula InChI=1S/C18H20N2O6/c1-5-26-18(22)15-11(3)19-10(2)14(17(21)25-4)16(15)12-7-6-8-13(9-12)20(23)24/h6-9,16,19H,5H2,1-4H3
Solution Type Neat
SMILES CCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC)C
IUPAC Name 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Industry Type Clinical Testing
Molecular Weight (g/mol) 360.36
Quantity 10 mg
Formula Weight 360.1321 g/mol