missing translation for 'onlineSavingsMsg'
Learn More

Nitrendipine

Product Code. 15473549
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473549 500 mg 500mg
15463549 100 mg 100mg
15453549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15473549 Supplier Thermo Scientific Alfa Aesar Supplier No. J60841.ME
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-type calcium channel blocker | CAS: 39562-70-4 | C18H20N2O6 | 360.366 g/mol

Nitrendipine modulates NMDA receptor channel function in mammalian neurons. Nitrendipine has been shown to inhibit neutrophil adhesion to vascular endothelium. It is used in the treatment of primary hypertension to decrease blood pressure. Nitrendipine is used alone or with an angiotensin-converting enzyme inhibitor, to treat hypertension, chronic stable angina pectoris, and Prinzmetal's variant angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO (17.5 mg/ml), methanol (15 mg/ml), ethyl acetate, and ethanol (25 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39562-70-4
Molecular Formula C18H20N2O6
Molecular Weight (g/mol) 360.366
MDL Number MFCD00082255
InChI Key PVHUJELLJLJGLN-UHFFFAOYSA-N
Synonym nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin
PubChem CID 4507
ChEBI CHEBI:7582
IUPAC Name 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C

Specifications

Color Yellow to Green
Quantity 500 mg
Merck Index 14,6576
Solubility Information Soluble in DMSO (17.5mg/ml),methanol (15mg/ml),ethyl acetate,and ethanol (25 mM). Insoluble in water.
Formula Weight 360.36
Physical Form Powder
Chemical Name or Material Nitrendipine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.