Learn More
Nitrendipine, Thermo Scientific™
L-type calcium channel blocker
195.00€ - 617.00€
Chemical Identifiers
| CAS | 39562-70-4 |
|---|---|
| Molecular Formula | C18H20N2O6 |
| Molecular Weight (g/mol) | 360.366 |
| MDL Number | MFCD00082255 |
| InChI Key | PVHUJELLJLJGLN-UHFFFAOYSA-N |
| Synonym | nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin |
| PubChem CID | 4507 |
| ChEBI | CHEBI:7582 |
| IUPAC Name | 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| SMILES | CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15463549
|
Thermo Scientific Alfa Aesar
J60841.MC |
100mg |
195.00€
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15473549
|
Thermo Scientific Alfa Aesar
J60841.ME |
500mg |
365.00€
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15453549
|
Thermo Scientific Alfa Aesar
J60841.03 |
1g |
617.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in DMSO (17.5 mg/ml), methanol (15 mg/ml), ethyl acetate, and ethanol (25 mM). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 39562-70-4 | |
| 360.366 | |
| PVHUJELLJLJGLN-UHFFFAOYSA-N | |
| 4507 | |
| 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| C18H20N2O6 | |
| MFCD00082255 | |
| nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin | |
| CHEBI:7582 | |
| CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C |
Specifications
| 39562-70-4 | |
| MFCD00082255 | |
| nitrendipine, bayotensin, baypress, nidrel, nitrendipinum, nitrendipino, bylotensin, nitrendipin, deiten, nitrepin | |
| PVHUJELLJLJGLN-UHFFFAOYSA-N | |
| 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate | |
| 4507 | |
| 360.36 |
| C18H20N2O6 | |
| 14,6576 | |
| Soluble in DMSO (17.5mg/ml),methanol (15mg/ml),ethyl acetate,and ethanol (25 mM). Insoluble in water. | |
| CCOC(=O)C1=C(NC(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)C | |
| 360.366 | |
| CHEBI:7582 | |
| Nitrendipine |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'einecsNumber' : 254-513-1
missing translation for 'rtecsNumber' : US5653000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.