missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 14% (w/w), Ricca Chemical

Product Code. 31940476
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940476 500 mL Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940476 Supplier Ricca Chemical Company Supplier No. R5323500500C
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Color Colorless to Yellow
CAS 7732-18-5, 7697-37-2
Odor Odorless to slightly pungent
Packaging Amber Glass Bottle
Assay Percent Range 14% (w/w)
Quantity 500 mL
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Percent Purity 14% W/W
Grade Laboratory
Chemical Name or Material Nitric Acid, Aqueous Solution
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.