missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 2% (w/w), Ricca Chemical

Product Code. 31940472
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940472 500 mL Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940472 Supplier Ricca Chemical Company Supplier No. R5315000500C
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Color Colorless to Yellow
CAS 7732-18-5, 7697-37-2
Odor Odorless to slightly pungent
Packaging Amber Glass Bottle
Assay Percent Range 2% (w/w)
Quantity 500 mL
Molecular Formula HNO3
Physical Form Liquid
MDL Number MFCD00011349
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Percent Purity 2% W/W
Grade Laboratory
Chemical Name or Material Nitric Acid, Aqueous Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.