Learn More
Chemsplash Multi-Task Nitrile Powder Free Disposable Gloves, Style 4010
Product Code. 30058218
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058218 Large Pack of 1000
30058198 Small Pack of 1000
30058274 Medium Pack of 1000
30058474 X-Large Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30058218 Supplier Chemsplash Supplier No. 4010.300.006
  • ONLINE EXCLUSIVE PRICE
    69.10€
    62.44€ / Pack of 1000
    Save 6.66€
    9% Off
View all promotions
Estimated Shipment: 23-11-2026
Add to basket
Aggiungi al carrello
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Premium nitrile gloves with textured fingertips meet medical grade standards and are food contact approved.
  • Food Contact Approved
  • Medical Grade
  • Ambidextrous
  • Rolled Cuff and Latex Free
  • PFAS Free
  • Touchscreen Compatible
  • CE and UKCA Approved
  • Thickness: 2.8 mil, Weight: 3.5 g
  • AQL: 1.5
  • Suitable for chemical splash protection and have some elasticated properties, allowing for ease of use and fit.
  • BRC-approved work areas and the Pharmaceutical Industry.
  • Free from PFAS, RoHS, POP, TSCA, REACH, or any Conflict Materials.
Specifica
ANSI Abrasion Rating ANSI 0
Certifications/Compliance (Europe) EN374-1: KPT, EN374-5: VIRUS, EN16523, EN455: 1/2/3 Medical Devices, EN1186: Food Grade, EN420
Chlorinated No
Applications Laboratory
Color Blue
Finger Thickness (English) 2.8 mil
Finger Thickness (Metric) 0.07 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.8 mil
Palm Thickness (Metric) 0.07 mm
Product Type Disposable Gloves
Size Large
Thickness (English) 2.8 mil
Thickness (Metric) 0.07 mm
EN 421 No
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Rolled
Lining Unlined
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.