missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene-d5, TRC

Product Code. 30478301
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478301 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478301 Supplier Toronto Research Chemicals Supplier No. TRCN49300210G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrobenzene D5
CAS 4165-60-0
Molecular Formula C6 2H5 N O2
Quantity 10 g
Synonym Nitrobenzene-D5, Nitrobenzene D5, Benzene-1,2,3,4,5-d5','6-nitro-, Benzene-d5','nitro- (6CI,7CI,8CI,9CI), 6-Nitrobenzene-1,2,3,4,5-d5, Nitrobenzene-d5, Pentadeuterionitrobenzene, Perdeuteronitrobenzene
InChI Formula InChI=1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])[N+](=O)[O-]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-nitrobenzene
Molecular Weight (g/mol) 128.14
Formula Weight 128.0634
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.