missing translation for 'onlineSavingsMsg'
Learn More

Nitrodenafil, TRC

Product Code. 30134481
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134481 5 g 5g
17912145 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134481 Supplier Toronto Research Chemicals Supplier No. TRCN4937705G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147676-99-1
Molecular Formula C17 H19 N5 O4
Molecular Weight (g/mol) 357.36
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-(2-ethoxy-5-nitrophenyl)-1,6-dihydro-1-methyl-3-propyl-, 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-(2-ethoxy-5-nitrophenyl)-1,4-dihydro-1-methyl-3-propyl- (9CI), 5-(2-Ethoxy-5-nitrophenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Nitrodenafil
IUPAC Name 5-(2-ethoxy-5-nitrophenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H19N5O4/c1-4-6-12-14-15(21(3)20-12)17(23)19-16(18-14)11-9-10(22(24)25)7-8-13(11)26-5-2/h7-9H,4-6H2,1-3H3,(H,18,19,23)
Formula Weight 357.1437
Percent Purity >95%
Chemical Name or Material Nitrodenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.