missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitroethane-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Nitroethane-D5 |
| CAS | 57817-88-6 |
| Molecular Formula | C2 D5 N O2 |
| Quantity | 0.5 g |
| Synonym | Ethane-d5, nitro- (9 CI), Nitroethane-d5, 1-Nitroethane-1,1,2,2,2-d5 |
| InChI Formula | InChI=1 S/C2H5NO2/c1-2-3(4)5/h2H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])[N+](=O)[O-] |
| IUPAC Name | 1,1,1,2,2-pentadeuterio-2-nitroethane |
| Molecular Weight (g/mol) | 80.0974 |
| Formula Weight | 80.0634 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction