missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin-13C3, TRC

Product Code. 18010420
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18010420 5 mg 5mg
18050410 1 mg 1mg
18082910 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18010420 Supplier Toronto Research Chemicals Supplier No. TRCN4938525MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217226-46-4
Molecular Formula 13C3 C5 H6 N4 O5
Molecular Weight (g/mol) 241.14
Synonym Labeled Nitrofurantoin, Nitrofurantoin-13C3
IUPAC Name 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]-(2,4,5-^{13}C_{3})1,3-diazolidine-2,4-dione
SMILES [O-][N+](=O)c1oc(\C=N\N2[13CH2][13C](=O)N[13C]2=O)cc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+/i4+1,6+1,8+1
Formula Weight 241.0439
Percent Purity >95%
Chemical Name or Material Nitrofurantoin 13C3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.