Learn More
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale
Product Code. 11862430
Change view
Click to view available options
Airflow Rate:
12 L/min.
120 mL/min.
200 mL/min.
6 mL/min.
60 mL/min.
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
11862430 12 L/min. Each
11852430 200 mL/min. Each
11892420 120 mL/min. Each
11862420 60 mL/min. Each
11802410 6 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11862430 Supplier Aalborg™ Supplier No. FT01416STVN
  • ONLINE EXCLUSIVE PRICE
    76.65€
    46.92€ / Pack of 1
    Save 29.73€
    38% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Nitrogen Direct Flow Tube
Width (Metric) 65 mm
Airflow Rate 12 L/min.