missing translation for 'onlineSavingsMsg'
Learn More

Nitrosoantipyrine, TRC

Product Code. 30240958
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240958 50 mg 50mg
30240952 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240958 Supplier Toronto Research Chemicals Supplier No. TRCN49195550MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885-11-0
Molecular Formula C11 H11 N3 O2
Molecular Weight (g/mol) 217.22
Synonym 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-4-nitroso-2-phenyl- (9CI, ACI), 1,2-Dihydro-1,5-dimethyl-4-nitroso-2-phenyl-3H-pyrazol-3-one (ACI), Antipyrine, 4-nitroso- (6CI, 7CI, 8CI), 1,2-Dihydro-2,3-dimethyl-4-nitroso-1-phenylpyrazol-5-one, 1,5-Dimethyl-4-nitroso-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, 1-Phenyl-2,3-dimethyl-4-nitroso-5-pyrazolone, 2,3-Dimethyl-4-nitroso-1-phenyl-5-pyrazolone, 4-Nitrosoantipyrin, 4-Nitrosoantipyrine, Nitrosoantipyrine, NSC 163932, NSC 75060
IUPAC Name 1,5-dimethyl-4-nitroso-2-phenylpyrazol-3-one
SMILES CN1N(C(=O)C(=C1C)N=O)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H11N3O2/c1-8-10(12-16)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,1-2H3
Formula Weight 217.0851
Chemical Name or Material Nitrosoantipyrine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.