missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrotyrosine Antibody (39B6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NB110-96877
This item is not returnable.
View return policy
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| Nitrotyrosine | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Microarray | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific. | |
| Store at -20°C. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 39B6 | |
| Unconjugated | |
| Nitrotyrosine | |
| Hybridoma line 39B6 | |
| 0.1 mg | |
| Primary | |
| Rat, Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction