missing translation for 'onlineSavingsMsg'
Learn More

Nitrovin Hydrochloride, TRC

Product Code. 30478657
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30478657 100 mg
30472392 25 mg
30471907 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478657 Supplier Toronto Research Chemicals Supplier No. TRCN494710100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-20-0
Molecular Formula C14 H12 N6 O6 . Cl H
Molecular Weight (g/mol) 396.74
Synonym Nitrovin hydrochloride, Hydrazinecarboximidamide, 2-[3-(5-nitro-2-furanyl)-1-[2-(5-nitro-2-furanyl)ethenyl]-2-propen-1-ylidene]-, hydrochloride (1:1), Guanidine, [[3-(5-nitro-2-furyl)-1-[2-(5-nitro-2-furyl)vinyl]allylidene]amino]-, monohydrochloride (8CI), Hydrazinecarboximidamide, 2-[3-(5-nitro-2-furanyl)-1-[2-(5-nitro-2-furanyl)ethenyl]-2-propenylidene]-, monohydrochloride (9CI), 1,5-Bis(5-nitro-2-furyl)penta-1,4-dien-3-one amidinohydrazone hydrochloride, IF 326, Panazon, Payzone, Payzone 250
IUPAC Name 2-[1,5-bis(5-nitrofuran-2-yl)penta-1,4-dien-3-ylideneamino]guanidine;hydrochloride
SMILES Cl.NC(=NN=C(C=Cc1oc(cc1)[N+](=O)[O-])C=Cc2oc(cc2)[N+](=O)[O-])N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H12N6O6.ClH/c15-14(16)18-17-9(1-3-10-5-7-12(25-10)19(21)22)2-4-11-6-8-13(26-11)20(23)24;/h1-8H,(H4,15,16,18);1H
Formula Weight 396.0585
Chemical Name or Material Nitrovin hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.