Learn More
Invitrogen™ MTHFR Recombinant Rabbit Monoclonal Antibody (6Q5C1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558126
Description
Immunogen sequence: RPIFWASRPK SYIYRTQEWD EFPNGRWGNS SSPAFGELKD YYLFYLKSKS PKEELLKMWG EELTSEESVF EVFVLYLSGE PNRNGHKVTC LPWNDEPLAA ETSLLKEELL RVNRQGILTI NSQPNINGKP SSDPIVGWGP SGGYVFQKAY LEFFTSRETA EALLQVLKKY ELRVNYHLVN VKGENITNAP ELQPNAVTWG IFPGREIIQP TVVDPVSFMF WKDEAFALWI ERWGKLYEEE SPSRTIIQYI HDNYFLVNLV DNDFPLDNCL WQVVEDTLEL LNRPTQNARE TEAP.
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
Specifications
| MTHFR | |
| Recombinant Monoclonal | |
| 1.31 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P42898, Q9WU20 | |
| MTHFR | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6Q5C1 | |
| Unconjugated | |
| MTHFR | |
| 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17769, 362657, 4524 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.