missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nizatidine, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Nizatidine |
| CAS | 76963-41-2 |
| Analyte or Component Names | Nizatidine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H21 N5 O2 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Nizatidine, N'-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N-methyl-2-nitro-1,1-ethenediamine, N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, Acinon, Axid, Bonacid, Calmaxid, Cronizat, Distaxid, Gastrax, LY 139037, Naxidine, Nizax, Nizaxid, Tazac, ZL 101, Zanizal |
| InChI Formula | InChI=1S/C12H21N5O2S2/c1-13-11(6-17(18)19)14-4-5-20-8-10-9-21-12(15-10)7-16(2)3/h6,9,13-14H,4-5,7-8H2,1-3H3 |
| Solution Type | Neat |
| SMILES | CNC(=C[N+](=O)[O-])NCCSCc1csc(CN(C)C)n1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction