missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539282
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5′ untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
Specifications
| NKp30 | |
| Polyclonal | |
| Unconjugated | |
| NCR3 | |
| 1C7, CD337, LY117, MALS, NKp30, NK-p30, activating NK-A1 receptor, activating natural killer receptor p30, lymphocyte antigen 117, natural killer cell p30-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 259197, 294251 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14931, Q8CFD9 | |
| NCR3 | |
| A synthetic peptide derived from the internal region of human NCR3 | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction