missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14763-1-AP-150UL
This item is not returnable.
View return policy
Description
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides.Specifications
| NLN | |
| Polyclonal | |
| Unconjugated | |
| NLN | |
| AGTBP, Angiotensin binding protein, KIAA1226, MEP, Microsomal endopeptidase, Mitochondrial oligopeptidase M, Neurolysin, mitochondrial, Neurotensin endopeptidase, NLN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117041, 57486, 75805 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42676, Q91YP2, Q9BYT8 | |
| NLN | |
| NLN Fusion Protein Ag6524 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction