missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NME2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149233
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
Specifications
| NME2 | |
| Polyclonal | |
| Unconjugated | |
| NME2 | |
| 2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 4831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P22392 | |
| NME2 | |
| E. coli-derived recombinant human NM23-H2, Ala2-Glu152. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction