Learn More
Invitrogen™ NME2 Recombinant Rabbit Monoclonal Antibody (24GB3730)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557124
This item is not returnable.
View return policy
Description
NME2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
Specifications
| NME2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NME2 | |
| 2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18103, 4831, 83782 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3730 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19804, P22392, Q01768 | |
| NME2 | |
| A synthesized peptide derived from human NME2 (aa 100-152). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |