missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NNMTi

Description
NNMTi is a nicotinamide N-methyltransferase (NNMT) inhibitor (IC50 = 1.2 μM). Promotes myoblast differentiation in C2C12 cells. Activates senescent muscle stem cells (muSCs) and improves regenerative capacity of aged skeletal muscle. Enhances proliferation, fusion and regenerative capacity of muSCs in aged mice.
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1-methylquinolinium iodide |
| CAS | 42464-96-0 |
| Quantity | 10 mg |
| Molecular Formula | C10H11IN2 |
| Purity | 97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?