missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NNMTi
GREENER_CHOICE

Product Code. 18798246
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18798246 50 mg 50mg
18706536 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18798246 Supplier Tocris Bioscience™ Supplier No. 6900/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nicotinamide N-methyltransferase (NNMT) inhibitor; promotes myoblast differentiation

NNMTi is a nicotinamide N-methyltransferase (NNMT) inhibitor (IC50 = 1.2 μM). Promotes myoblast differentiation in C2C12 cells. Activates senescent muscle stem cells (muSCs) and improves regenerative capacity of aged skeletal muscle. Enhances proliferation, fusion and regenerative capacity of muSCs in aged mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-1-methylquinolinium iodide
CAS 42464-96-0
Quantity 50 mg
Molecular Formula C10H11IN2
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.