Learn More
NNT Mouse, anti-Human, CoraLite Plus 488, Clone: 1F5H6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68194-20UL
Description
The NNT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NNT. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The NNT antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NNT | |
| 1F5H6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig | |
| 155 kDa | |
| IgG2b |
| Immunofluorescence | |
| 1000 μg/mL | |
| NNT | |
| 20 μL | |
| 23530 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |