Learn More
NNT Mouse, anti-Human, CoraLite Plus 488, Clone: 1F5H6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68194-20UL
 View more versions of this product
Product Code. 18056185
  • 145.00€ / 20µL
Description
The NNT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NNT. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The NNT antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
NNT
1F5H6
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat, Rabbit, Pig
155 kDa
IgG2b
Immunofluorescence
1000 μg/mL
NNT
20 μL
23530
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.