missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nogo Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577029
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.
Specifications
| Nogo Receptor | |
| Polyclonal | |
| Unconjugated | |
| RTN4R | |
| HGNC:1860; NgR; Ngr1; nogo 66 receptor; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; reticulon-4 receptor; Rtn4r; UNQ330/PRO526 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113912, 65078, 65079 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99M75, Q99PI8, Q9BZR6 | |
| RTN4R | |
| Recombinant full length Human Nogo Receptor. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction