missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Non-Metals Standard: As, B, P, S, Se, Si, Te @ 100 μg/mL in 5% HCl, tr. HF, VHG™
Description
Non-Metals Standard: As, B, P, S, Se, Si, Te @ 100 μg/mL in 5% HCl, tr. HF
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Selenium/Boron/Tellurium/Sulfur/Silicon/Arsenic |
| CAS | 7723-14-0, 7782-49-2, 7440-42-8, 13494-80-9, 7704-34-9, 7440-21-3 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Phosphorus/Selenium/Boron/Tellurium/Sulfur/Silicon/Arsenic |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Phosphorus 99.96 μg/mL/Selenium 99.97 μg/mL/Boron 99.99 μg/mL/Tellurium 99.99 μg/mL/Sulfur 100 μg/mL/Silicon 100.1 μg/mL/Arsenic 100.2 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?