missing translation for 'onlineSavingsMsg'
Learn More

Nonanoyl Chloride, TRC

Product Code. 30483562
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483562 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483562 Supplier Toronto Research Chemicals Supplier No. TRCN64937510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nonanoyl Chloride
CAS 764-85-2
Molecular Formula C9H17ClO
Quantity 10 g
Synonym NSC 9829, Nonanoic Acid Chloride, Pelargonic Acid Chloride, Pelargonoyl Chloride, Pelargonyl Chloride, n-Nonanoyl Chloride
InChI Formula InChI=1S/C9H17ClO/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3
SMILES CCCCCCCCC(=O)Cl
IUPAC Name nonanoyl chloride
Molecular Weight (g/mol) 176.68
Formula Weight 176.0968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.