missing translation for 'onlineSavingsMsg'
Learn More

Nor Harmane, TRC

Product Code. 30135396
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135396 500 mg 500mg
30133934 1 g 1g
18010050 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135396 Supplier Toronto Research Chemicals Supplier No. TRCN700000500MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244-63-3
Molecular Formula C11 H8 N2
Molecular Weight (g/mol) 168.19
Synonym 2,9-Diazafluorene, 2-Azacarbazole, Carbazoline, NSC 84417, Norharman, Norharmane, β-Carboline, 9H-Pyrido[3,4-b]indole
IUPAC Name 9H-pyrido[3,4-b]indole
SMILES c1ccc2c(c1)[nH]c3cnccc23

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H8N2/c1-2-4-10-8(3-1)9-5-6-12-7-11(9)13-10/h1-7,13H
Formula Weight 168.0687
Percent Purity >95%
Chemical Name or Material Norharman
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.