missing translation for 'onlineSavingsMsg'
Learn More

Nor Scopolamine, TRC

Product Code. 30040991
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040991 50 mg 50mg
30041205 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040991 Supplier Toronto Research Chemicals Supplier No. TRCN82588050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4684-28-0
Molecular Formula C16 H19 N O4
Molecular Weight (g/mol) 289.33
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, (1α,2β,4β,5α,7β)-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, (αS)-, 1αH,5αH-Nortropan-3α-ol, 6β,7β-epoxy-, (-)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, [7(S)-(1α,2β,4β,5α,7β)]-, Norscopolamine (7CI), Norhyoscine
SMILES OC[C@@H](C(=O)O[C@H]1C[C@@H]2N[C@H](C1)[C@H]3O[C@@H]23)c4ccccc4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H19NO4/c18-8-11(9-4-2-1-3-5-9)16(19)20-10-6-12-14-15(21-14)13(7-10)17-12/h1-5,10-15,17-18H,6-8H2/t10-,11-,12-,13+,14-,15+/m1/s1
Formula Weight 289.1314
Percent Purity >95%
Chemical Name or Material Norhyoscine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.