missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nordiphenhydramine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Nordiphenhydramine Hydrochloride |
| CAS | 53499-40-4 |
| Analyte or Component Names | Nordiphenhydramine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride (Nordiphenhydramine Hydrochloride), Dimenhydrinate Imp. F (EP), Diphenhydramine Hydrochloride Imp. A (EP), Diphenhydramine Imp. A (EP), Diphenhydramine USP Related Compound A, Diphenhydramine USP RC A, 2-(Diphenylmethoxy)-N-methylethanamine Hydrochloride, Nordiphenhydramine Hydrochloride, Dimenhydrinate Imp. F (EP) as Hydrochloride, Diphenhydramine Imp. A Hydrochloride, Diphenhydramine Hydrochloride Imp. A (EP) as Hydrochloride, Diphenhydramine Related Compound A as Hydrochloride, Dimenhydrinate Impurity F as Hydrochloride, Diphenhydramine Hydrochloride Impurity A as Hydrochloride, Diphenhydramine Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C16H19NO.ClH/c1-17-12-13-18-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15;/h2-11,16-17H,12-13H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CNCCOC(c1ccccc1)c2ccccc2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?