missing translation for 'onlineSavingsMsg'
Learn More

Norepinephrine Sulfonic Acid, TRC

Product Code. 30041029
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041029 50 mg 50mg
30041271 100 mg 100mg
30040759 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041029 Supplier Toronto Research Chemicals Supplier No. TRCN67453050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24159-36-2
Molecular Formula C8 H11 N O5 S
Molecular Weight (g/mol) 233.24
Synonym α-(Aminomethyl)-3,4-dihydroxy-benzenemethanesulfonic Acid
IUPAC Name 2-amino-1-(3,4-dihydroxyphenyl)ethanesulfonic acid
SMILES NCC(c1ccc(O)c(O)c1)S(=O)(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11NO5S/c9-4-8(15(12,13)14)5-1-2-6(10)7(11)3-5/h1-3,8,10-11H,4,9H2,(H,12,13,14)
Formula Weight 233.0358
Percent Purity >95%
Chemical Name or Material Norepinephrine sulfonic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.