missing translation for 'onlineSavingsMsg'
Learn More

Norgestrel, TRC

Product Code. 30138347
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138347 250 mg 250mg
30136209 1 g 1g
30041261 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138347 Supplier Toronto Research Chemicals Supplier No. TRCN689500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6533-00-2
Molecular Formula C21 H28 O2
Molecular Weight (g/mol) 312.45
Synonym Norgestrel, rac-13-Ethyl-17-hydroxy-18,19-dinor-17a-pregn-4-en-20-yn-3-one, 18,19-Dinor-17α-pregn-4-en-20-yn-3-one, 13-ethyl-17-hydroxy-, (±)- (8CI), (±)-13-Ethyl-17α-ethynyl-17-hydroxygon-4-en-3-one, (±)-17α-Ethynyl-13-ethyl-17-hydroxygon-4-en-3-one, (±)-17α-Ethynyl-13β-ethylgon-4-en-17β-ol-3-one, (±)-Norgestrel, 13-Ethyl-17α-ethynyl-17β-hydroxy-4-gonen-3-one, DL-Norgestrel, FH 122A, Methylnorethindrone, Monovar, Neogest, Norgestrel, Ovrette, Postinor, SH 70850, SH 850, Wy 3707, dl-11β-Ethyl-17α-ethynyl-17β-hydroxygon-4-en-3-one, dl-13-β-Ethyl-17-α-ethynyl-17-β-hydroxygon-4-en-3-one, dl-Norgestrel, α-Norgestrel
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
Formula Weight 312.2089
Percent Purity >95%
Chemical Name or Material Norgestrel
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.