missing translation for 'onlineSavingsMsg'
Learn More

Normal Goat IgG, Alexa Fluor 488, Negative Control, R&D Systems™

Product Code. 18154031
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18154031 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18154031 Supplier R&D Systems Supplier No. IC108G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Control 5 uL/10^6 cells
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Immunogen NA
Purification Method Protein A or G purified
Quantity 200 Tests
Gene ID (Entrez) 3500
Test Specificity Serum was obtained from naive (non-immunized) goats.
Target Species Goat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.