missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-3 Antibody (603532) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1559X
498.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Notch-3 Monoclonal antibody specifically detects Notch-3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Notch-3 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
603532 | |
Flow Cytometry, CyTOF-ready | |
CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Notch-3, Ala40-Glu467, Accession # Q9UM47 | |
0.1 mL | |
Cancer | |
4854 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction