Learn More
NOTCH1 Monoclonal Antibody (OTI2E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808308S
Description
NOTCH1 Monoclonal antibody specifically detects NOTCH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NOTCH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75473 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8549 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI2E7 | |
| Unconjugated | |
| Lgr5 | |
| 9930111A19Rik; AOS5; AOVD1; Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); hN1; lin-12; major type A protein; Mis6; Motch; Motch A; mT14; N1; neurogenic locus notch homolog protein 1; NEXT; NICD; NOTCH; notch 1; Notch 1 extracellular truncation; Notch 1 intracellular domain; Notch gene homolog 1; Notch homolog 1, translocation-associated; notch receptor 1; Notch1; p300; RP23-306D20.12; RP23-306D20.12-001; Tan1; Translocation-associated notch protein TAN-1; transmembrane receptor Notch1 | |
| Human recombinant protein fragment corresponding to amino acids 22-543 of human LGR5 produced in HEK293 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.