Learn More
Invitrogen™ Empty Gel Cassette Combs, mini, 1.5 mm, 10 well
Product Code. 10462142
Change view
Click to view available options
Quantity:
25⁄pkg
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10462142 25⁄pkg Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10462142 Supplier Invitrogen™ Supplier No. NC3510
Compatible with mini-size Empty Gel Cassettes of the same thickness (NC2010, 1.0mm; NC2015, 1.5mm)
These combs are for use when pouring special gel types and are available in two thicknesses (1.0mm and 1.5mm) and seven well formats (1 well, 2 well, 5 well, 10 well, 12 well, 15 well, and 2D well).
Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Includes 1.5 mm x 10 well combs (25⁄pack).
For Use With (Equipment) Universal
Gel Size Mini
Product Line Novex
Type 10-well Empty Gel Cassette Comb
Quantity 25⁄pkg
Shipping Condition Room Temperature
Thickness (Metric) 1.5 mm