missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminex™ Intracellular Protein Buffer Reagent Kit

Product Code. 10486473
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10486473 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10486473 Supplier Invitrogen™ Supplier No. LHB0002

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Includes

Diluents, SAV concentrate, buffers, wash solution, filter plate and complete protocol to be followed in conjunction with the selected Human, Mouse and Rat Intracellular Singleplex Bead Kits.

Requires

Human, Mouse or Rat Intracellular Singleplex Bead Kits

Use with Human, Mouse and Rat Intracellular Singleplex Bead Kits for in vitro quantitative determination of analytes in cell lysates and tissue homogenates

The Intracellular Protein Buffer Reagent Kit is comprised of the reagents intended for use with Human, Mouse and Rat Intracellular Singleplex Bead Kits. Singleplex Bead Kits have been developed to maximize flexibility in experimental design, permitting the measurement of one or multiple analytes in panels designed by the researcher. These reagents are suitable for use with the Luminex 100™/200™ and the FLEXMAP 3D™ systems.

Luminex xMAP™ Technology—Effective and Proven Analytical Tool
The Human, Mouse and Rat Intracellular Singleplex Bead Kits used in conjunction with the Intracellular Protein Buffer Reagent Kit are based on xMAP™ technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Polystyrene microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. At the completion of the sandwich immunoassay, beads can be read using theLuminex 100/200 or FLEXMAP 3D detection systems, instruments based on flow cytometry, two lasers, real-time digital signal processing, and xPONENT™ software to distinguish bead color (analyte) and assay signal strength (PE) fluorescence intensity. The Human, Mouse and Rat Intracellular Singleplex Bead Kits that are used with the Intracellular Protein Buffer Reagent Kit have been validated using the rigorous criterion that distinguishes Luminex assays from other assays.

  • Accurate, reproducible, and sensitive quantitation of multiple proteins
  • Fully qualified antibodies permit specificity and sensitivity
  • Performs assay and analyzes data typically in less than one day

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 Tests
Species Human, Mouse, Rat
Sample Type Cell Extracts

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.