Description
- Autoclavable
- Flexible and soft
- Suitable for hose pumps
- Resistant to mold, water, radiation and light effects
- Excellent compatibility with sterile solutions
- Applications include pharmaceutical and biotechnology
- Not recommended for concentrated solvents, oils, diluted acids and alkalis
Specifications
Specifications
| Certifications/Compliance | FDA, BFR and WRAS approved |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 8 mm |
| Material | Silicone |
| Diameter (Metric) Outer | 14 mm |
| Overall Length (Metric) | 5 m |
| For Use With (Application) | Ideal for pharmaceutical, biotechnology, sterile fluids transfers |
| Min. Operating Temperature (Metric) | -40°C |
| Color | Translucent |
| Flexibility | Flexible |
| Show More |