missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CXCR1/IL-8RA Overexpression Lysate

Product Code. 18215321 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215321 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215321 Supplier Novus Biologicals™ Supplier No. NBL111968

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The CXCR1/IL-8RA Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC2719 and based on accession number NM_634. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens interleukin 8 receptor, alpha (IL8RA), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Research Category Cytokine Research, GPCR, Signal Transduction
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.