missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 12-261 of Human ERAB The Recombinant Human ERAB Protein is derived from E. coli. The Recombinant Human ERAB Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_004484 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT, 100mM NaCl |
| Gene ID (Entrez) | 3028 |
| Molecular Weight (g/mol) | 28.1kDa |
| Name | ERAB Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MVAVITGGAS GLGLATAERL VGQGASAVLL DLPNSGGEAQ AKKLGNNCVF APADVTSEKD VQTALALAKG KFGRVDVAVN AGIAVASKT YNLKKGQTHT LEDFQRVLDV NLMGTFNVIR LVAGEMGQNE PDQGGQRGVI INTASVAAFE GQVGQAAYSA SKGGIVGMTL PIARDLAPIG IRVMTIAPGL FGTPLLTSLP EKVCNFLASQ VPFPSRLGDP AEYAHLVQAI IENPFLNGEV IRLDGAIRMQ P |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?