missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human P311 His Protein

Product Code. 18197471 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
Unit Size:
0.02mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18197471 0.02 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18197471 Supplier Novus Biologicals™ Supplier No. NBP223281

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-68 of Human P311 The Recombinant Human P311 Protein is derived from E. coli. The Recombinant Human P311 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004763
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
Gene ID (Entrez) 9315
Molecular Weight (g/mol) 10.3kDa
Name P311 Protein
Purification Method SDS-PAGE
Quantity 0.02 mg
Source E. Coli
Immunogen NREP, 1-68aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMVYYPEL FVWVSQEPFP NKDMEGRLPK GRLPVPKEVN RKKNDETNAA SLTPLGSSEL RSPRISYLHF F
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.