Learn More
Novus Biologicals™ Recombinant Human Renin His Protein
Product Code. 18760314 Shop All Bio Techne Products
Product Code. 18760314 Supplier Novus Biologicals™ Supplier No. NBP223408
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 67-406 of Human Renin The Recombinant Human Renin Protein is derived from E. coli. The Recombinant Human Renin Protein has been validated for the following applications: SDS-Page.