missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human TEN1 His Protein

Product Code. 18148571 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18148571 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18148571 Supplier Novus Biologicals™ Supplier No. NBP222855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-123 of Human TEN1 The Recombinant Human TEN1 Protein is derived from E. coli. The Recombinant Human TEN1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001106795
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 0.15 M NaCl, 10% glycerol, 1mM DTT
Gene ID (Entrez) 100134934
Molecular Weight (g/mol) 16.2kDa
Name TEN1 Protein
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. Coli
Immunogen TEN1, 1-123aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMMLPKPG TYYLPWEVSA GQVPDGSTLR TFGRLCLYDM IQSRVTLMAQ HGSDQHQVLV CTKLVEPFHA QVGSLYIVLG ELQHQQDRGS VVKARVLTCV EGMNLPLLEQ AIREQRLYKQ ERGGSQ
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.