missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 1 Kit |
| Target | QuikChIP |
| Kit Contents | Wash Buffer A- 25mL, Wash Buffer B-25mL, Wash Buffer C- 25mL, Wash Buffer D- 50mL, SDS Lysis Buffer-15mL, 10X Glycine- 25mL, 10X PBS-55mL, ChIP Dilution Buffer- 50mL, Salmon Sperm DNA/ Protein A/G Agarose- 2 x 1.75mL, RNAse-10 mg/ml 60μL, 100X PMSF-0.75mL, 100X PIC-0.75mL, Proteinase K (10 mg/ml)-60μL, 20% SDS-625μL, 1M NaHCO3-1.25mL, 0.5M EDTA-250μL, 5M NaCl -500μL, 1M Tris-HCl- pH 6.5-500μL |
| For Use With (Application) | Chromatin Immunoprecipitation |
| Product Type | Kit (ChIP) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?