missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C1A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06431PE
This item is not returnable.
View return policy
Description
NT5C1A Polyclonal antibody specifically detects NT5C1A in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NT5C1A | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 5'-nucleotidase, cytosolic IA, AMP-specific 5'-NT, CN1, cN1A, CN-I, cN-IA, cytosolic 5' nucleotidase, type 1A, cytosolic 5'-nucleotidase 1A, Cytosolic 5'-nucleotidase IA, EC 3.1.3.5, MGC119199, MGC119201 | |
| Produced in rabbits immunized with E. coli-derived Human NT5C1A fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84618 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction