missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nox4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99108-50ul
This item is not returnable.
View return policy
Description
Nox4 Polyclonal antibody specifically detects Nox4 in Human samples. It is validated for Immunoprecipitation
Specifications
| Nox4 | |
| Polyclonal | |
| Immunoprecipitation 1-4 μg/mg of lysate | |
| EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nox4. | |
| 50 μg | |
| Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
| 50507 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction