Learn More
NPHP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22026-1-AP-20UL
This item is not returnable.
View return policy
Description
Inhibition of the nuclear export of poly(A)-containing mRNAs caused by the influenza A virus NS1 protein requires its effector domain. The NS1 effector domain functionally interacts with the cellular 30 kDa subunit of cleavage and polyadenylation specific factor 4, an essential component of the 3′ end processing machinery of cellular pre-mRNAs. In influenza virus-infected cells, the NS1 protein is physically associated with cleavage and polyadenylation specific factor 4, 30kD subunit. Binding of the NS1 protein to the 30 kDa protein in vitro prevents CPSF binding to the RNA substrate and inhibits 3′ end cleavage and polyadenylation of host pre-mRNAs. Thus the NS1 protein selectively inhibits the nuclear export of cellular, and not viral, mRNAs. Multiple alternatively spliced transcript variants that encode different isoforms have been described for this gene.Specifications
| NPHP3 | |
| Polyclonal | |
| Unconjugated | |
| NPHP3 | |
| KIAA2000, MKS7, Nephrocystin 3, NPHP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27031, 74025 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TNH6, Q7Z494 | |
| Nphp3 | |
| NPHP3 Fusion Protein Ag16888 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |